missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Dnak SBD C-terminus Protein
A cDNA sequence encoding the Dnak SBD C-terminus was constructed and used to recombinantly synthesize the protein.
£99.15 - £1100.00
Specifications
Name | Dnak SBD C-terminus Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15948048
|
enQuireBio™
QP11688-20UG |
20 μg |
£99.15
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15928048
|
enQuireBio™
QP11688-100UG |
100 μg |
£155.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938048
|
enQuireBio™
QP11688-1MG |
1 mg |
£1100.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Dnak SBD C-terminus Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
The DnaK protein contains 25mM Tris-HCl, pH 7.5, 100mM NaCl, 5mM DTT and 10% Glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MDVKDVLLLD VTPLSLGIET MGGVMTTLIA KNTTIPTKHS QVFSTAEDNQ SAVTIHVLQGERKRAADNKS LGQFNLDGIN PAPRGMPQIE VTFDIDADGI LHVSAKDKNS GKEQKITIKA SSGLNEDEIQ KMVRDAEANA EADRKFEELV QTRNQGDHLL HSTRKQVEEA GDKLPADDKTAIESALTALE TALKGEDKAA IEAKMQELAQ VSQKLMEIAQ QQHAQQQTAG ADASANNAKD DDVVDAEFEE VKDKK | |
Greater than 95.0% as determined by SDS-PAGE. |