missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Actin Protein
A cDNA sequence encoding the Actin was constructed and used to recombinantly synthesize the protein.
£99.15 - £1655.00
Specifications
Name | Actin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Other |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15965668
|
enQuireBio™
QP10951-10UG |
10 μg |
£99.15
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15985668
|
enQuireBio™
QP10951-50UG |
50 μg |
£155.00
50µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975668
|
enQuireBio™
QP10951-1MG |
1 mg |
£1655.00
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Actin Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 98.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Non-species Specific | |
The protein was lyophilized from a 1 mg/ml solution containing 10mM Tris/HCl buffer pH 8.0, 0.2mM CaCl2, 0.2mM ATP, 1mM DTT and 0.5% (w/v) SDS. |