missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Actin Protein
A cDNA sequence encoding the Actin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10951-1mg
Additional Details : Weight : 0.01000kg
Specifications
Actin Protein | |
Research Use Only | |
Recombinant Protein | |
Non-species Specific | |
The protein was lyophilized from a 1 mg/ml solution containing 10mM Tris/HCl buffer pH 8.0, 0.2mM CaCl2, 0.2mM ATP, 1mM DTT and 0.5% (w/v) SDS. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Greater than 98.0% as determined by SDS-PAGE. |