missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse TRANCE/RANK L/TNFSF11 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£291.57
Specifications
Gene ID (Entrez) | 21943 |
---|---|
Molecular Weight (g/mol) | 28 kDa |
Name | TRANCE/TNFSF11/RANK L |
Quantity | 10μg |
Source | Mouse myeloma cell line,NS0-derived mouse TRANCE/TNFSF11/RANK L protein Arg72-Asp316,with an N-terminal 6-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18146330
|
R&D Systems™
462-TR-010 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£291.57
10µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18395007
|
R&D Systems™
462-TR-010/CF |
£291.57
10µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse TRANCE/RANK L/TNFSF11 Protein is derived from NS0. The Recombinant Mouse TRANCE/RANK L/TNFSF11 Protein has been validated for the following applications: Bioactivity.Specifications
21943 | |
TRANCE/TNFSF11/RANK L | |
Mouse myeloma cell line,NS0-derived mouse TRANCE/TNFSF11/RANK L protein Arg72-Asp316,with an N-terminal 6-His tag | |
Recombinant |
28 kDa | |
10μg | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |