missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse TRANCE/RANK L/TNFSF11 Protein

Product Code. 18395007
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Supplied as a 0.2μm filtered solution in PBS.
Unit Size:
10µg
This item is not returnable. View return policy

Product Code. 18395007

Brand: R&D Systems™ 462TR010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse TRANCE/RANK L/TNFSF11 Protein is derived from NS0. The Recombinant Mouse TRANCE/RANK L/TNFSF11 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 21943
Molecular Weight (g/mol) 28 kDa
Name TRANCE/TNFSF11/RANK L
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived mouse TRANCE/TNFSF11/RANK L protein Arg72-Asp316,with an N-terminal 6-His tag
Storage Requirements 3 months from date of receipt, 2 to 8° C as supplied.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.