missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse TNF-alpha (aa 84-235) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£209.50 - £571.26
Specifications
Gene ID (Entrez) | 21926 |
---|---|
Molecular Weight (g/mol) | 17 kDa (monomer) |
Name | TNF-alpha aa 84-235 |
Source | E. coli-derived mouse TNF-alpha protein Ser84-Leu235 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Quantity | Price | Quantity & Availability | ||||
18108340
|
R&D Systems™
410-TRNC-010 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 10μg |
£209.50
10µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18148150
|
R&D Systems™
410-TRNC-010/CF |
Lyophilized from a 0.2μm filtered solution in PBS. | 10μg |
£209.50
10µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18168060
|
R&D Systems™
410-TRNC-050 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | 50μg |
£571.26
50µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18138630
|
R&D Systems™
410-TRNC-050/CF |
Lyophilized from a 0.2μm filtered solution in PBS. | 50μg |
£571.26
50µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
The Recombinant Mouse TNF-alpha (aa 84-235) Protein is derived from E. coli. The Recombinant Mouse TNF-alpha (aa 84-235) Protein has been validated for the following applications: Bioactivity.Specifications
21926 | |
TNF-alpha aa 84-235 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
17 kDa (monomer) | |
E. coli-derived mouse TNF-alpha protein Ser84-Leu235 | |
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
Recombinant |