missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse TNF-alpha (aa 84-235) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 410-TRNC-050
Additional Details : Weight : 0.00970kg
Description
The Recombinant Mouse TNF-alpha (aa 84-235) Protein is derived from E. coli. The Recombinant Mouse TNF-alpha (aa 84-235) Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
17 kDa (monomer) | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
21926 | |
TNF-alpha aa 84-235 | |
E. coli-derived mouse TNF-alpha protein Ser84-Leu235 | |
APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha | |
Recombinant |