missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Serine protease inhibitor Kazal-type 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Serine protease inhibitor Kazal-type 1, was expressed in the hosts and tags indicated.
£223.39 - £2410.48
Specifications
Accession Number | NP_033284.1 |
---|---|
Gene ID (Entrez) | 20730 |
Name | Serine protease inhibitor Kazal-type 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048550
|
enQuireBio™
QP7376-YE-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088550
|
enQuireBio™
QP7376-YE-500UG |
500 μg |
£1600.00
500µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058550
|
enQuireBio™
QP7376-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078550
|
enQuireBio™
QP7376-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098550
|
enQuireBio™
QP7376-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068550
|
enQuireBio™
QP7376-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_033284.1 | |
Serine protease inhibitor Kazal-type 1 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
20730 | |
Research Use Only | |
Spink3 | |
Recombinant Protein | |
Yeast | |
AKVTGKEASC HDAVAGCPRI YDPVCGTDGI TYANECVLCF ENRKRIEPVL IRKGGPC | |
Greater than 80% as determined by SDS-PAGE. |