missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Retnlb Protein
Recombinant Protein
£80.80 - £3085.00
Specifications
Accession Number | NP_076370.3 |
---|---|
Gene ID (Entrez) | 57263 |
Name | Retnlb Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15946209
|
enQuireBio™
QP5475-5UG |
5 μg |
£80.80
5µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926209
|
enQuireBio™
QP5475-25UG |
25 μg |
£191.00
25µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15906209
|
enQuireBio™
QP5475-100UG |
100 μg |
£476.00
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936209
|
enQuireBio™
QP5475-500UG |
500 μg |
£1890.00
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15916209
|
enQuireBio™
QP5475-1MG |
1 mg |
£3085.00
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_076370.3 | |
Retnlb Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MQCSFESLVD QRIKEALSRQ EPKTISCTSV TSSGRLASCP AGMVVTGCAC GYGCGSWDIR NGNTCHCQCS VMDWASARCC RMA | |
Greater than 95% as determined by SDS-PAGE. |
57263 | |
Research Use Only | |
Retnlb | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |