missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Retnlb Protein

Recombinant Protein

Brand:  enQuireBio™ QP5475-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15946209

  • £77.85 / 5µg
Estimated Shipment: 24-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_076370.3
Retnlb Protein
5 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MQCSFESLVD QRIKEALSRQ EPKTISCTSV TSSGRLASCP AGMVVTGCAC GYGCGSWDIR NGNTCHCQCS VMDWASARCC RMA
57263
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Retnlb
Mouse
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
Videos
SDS
Documents
Certificates
Special Offers

Special Offers