missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Ret Fc Chimera Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
£345.56
Specifications
Gene ID (Entrez) | 19713 |
---|---|
Molecular Weight (g/mol) | 95 kDa (monomer) |
Name | Ret Fc Chimera |
Quantity | 100μg |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18166150
|
R&D Systems™
482-RT-100 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£345.56
100µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18197350
|
R&D Systems™
482-RT-100/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£345.56
100µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse Ret Fc Chimera Protein is derived from NS0. The Recombinant Mouse Ret Fc Chimera Protein has been validated for the following applications: Binding Activity.Specifications
19713 | |
Ret Fc Chimera | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
95 kDa (monomer) | |
100μg | |
Disulfide-linked homodimer | |
Recombinant |