missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Ret Fc Chimera Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Brand:  R&D Systems™ 482-RT-100/CF

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18197350

  • £345.56 / 100µg
Estimated Shipment: 06-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse Ret Fc Chimera Protein is derived from NS0. The Recombinant Mouse Ret Fc Chimera Protein has been validated for the following applications: Binding Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
95 kDa (monomer)
100μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
19713
Ret Fc Chimera
Mouse myeloma cell line,NS0-derived mouse Ret protein, (N-terminus) Mouse cRet Leu29-Arg637 (Phe174Ser) Accession &Num; P35546.2 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Disulfide-linked homodimer
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers