missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Protein ABHD11 Protein
A DNA sequence encoding the Mus musculus (Mouse) Protein ABHD11, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7815-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_660250.1 | |
Protein ABHD11 Protein | |
Research Use Only | |
Abhd11 | |
Recombinant Protein | |
Yeast | |
MLRWARAWRV PRGVLGASSP RRLAVPVTFC SSRSSGQENA DLRPLPLSYN LLDGDATLPA IVFLHGLFGS KTNFNSLAKA MVQRTGRRVL TVDARNHGDS PHSPDASYEA MSQDLQGLLP QLGLVPCVLV GHSMGGKTAM LLALQRPDVV ERLVVVDISP VGTTPGSHIG AFIAAMKAVE IPEKVPHSQA RKLADKQLSS VVKEAGIRQF LLTNLVEVGG RFSWRLNLDT LAQHLDKIMT FPQQREPYSG PTLFLLGGNS TYVQPSHHSE IRRLFPQAQI QTVPNAGHWV HSDKPQDFMD AVTSFLA | |
Greater than 90% as determined by SDS-PAGE. |
68758 | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |