missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Protein ABHD11 Protein
A DNA sequence encoding the Mus musculus (Mouse) Protein ABHD11, was expressed in the hosts and tags indicated.
£223.39 - £2410.48
Specifications
Accession Number | NP_660250.1 |
---|---|
Gene ID (Entrez) | 68758 |
Name | Protein ABHD11 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16050961
|
enQuireBio™
QP7815-YE-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16090961
|
enQuireBio™
QP7815-YE-500UG |
500 μg |
£1600.00
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060961
|
enQuireBio™
QP7815-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080961
|
enQuireBio™
QP7815-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16000971
|
enQuireBio™
QP7815-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070961
|
enQuireBio™
QP7815-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_660250.1 | |
Protein ABHD11 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
68758 | |
Research Use Only | |
Abhd11 | |
Recombinant Protein | |
Yeast | |
MLRWARAWRV PRGVLGASSP RRLAVPVTFC SSRSSGQENA DLRPLPLSYN LLDGDATLPA IVFLHGLFGS KTNFNSLAKA MVQRTGRRVL TVDARNHGDS PHSPDASYEA MSQDLQGLLP QLGLVPCVLV GHSMGGKTAM LLALQRPDVV ERLVVVDISP VGTTPGSHIG AFIAAMKAVE IPEKVPHSQA RKLADKQLSS VVKEAGIRQF LLTNLVEVGG RFSWRLNLDT LAQHLDKIMT FPQQREPYSG PTLFLLGGNS TYVQPSHHSE IRRLFPQAQI QTVPNAGHWV HSDKPQDFMD AVTSFLA | |
Greater than 90% as determined by SDS-PAGE. |