missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Protein ABHD11 Protein

A DNA sequence encoding the Mus musculus (Mouse) Protein ABHD11, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7815-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16050961

  • £560.00 / 100µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_660250.1
Protein ABHD11 Protein
Research Use Only
Abhd11
Recombinant Protein
Yeast
MLRWARAWRV PRGVLGASSP RRLAVPVTFC SSRSSGQENA DLRPLPLSYN LLDGDATLPA IVFLHGLFGS KTNFNSLAKA MVQRTGRRVL TVDARNHGDS PHSPDASYEA MSQDLQGLLP QLGLVPCVLV GHSMGGKTAM LLALQRPDVV ERLVVVDISP VGTTPGSHIG AFIAAMKAVE IPEKVPHSQA RKLADKQLSS VVKEAGIRQF LLTNLVEVGG RFSWRLNLDT LAQHLDKIMT FPQQREPYSG PTLFLLGGNS TYVQPSHHSE IRRLFPQAQI QTVPNAGHWV HSDKPQDFMD AVTSFLA
Greater than 90% as determined by SDS-PAGE.
68758
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers