missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PRDX1 Protein
A cDNA sequence encoding the PRDX1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Gene ID (Entrez) | 18477 |
---|---|
Name | PRDX1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Prdx1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15975318
|
enQuireBio™
QP10837-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965318
|
enQuireBio™
QP10837-10UG |
10 μg |
£160.00
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955318
|
enQuireBio™
QP10837-100UG |
100 μg |
£1320.00
100µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
18477 | |
Research Use Only | |
Prdx1 | |
Recombinant Protein | |
Baculovirus | |
MSSGNAKIGY PAPNFKATAV MPDGQFKDIS LSEYKGKYVV FFFYPLDFTF VCPTEIIAFS DRADEFKKLN CQVIGASVDS HFCHLAWINT PKKQGGLGPM NIPLISDPKR TIAQDYGVLK ADEGISFRGL FIIDDKGILR QITINDLPVG RSVDEIIRLV QAFQFTDKHG EVCPAGWKPG SDTIKPDVNK SKEYFSKQKL EHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
PRDX1 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is >2,500 pmol/min/ug. Enzymatic activity is defined as the amount of hydroperoxide that 1ug of enzyme can reduce at 25C for minute. | |
Mouse | |
His | |
PRDX1 protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |