missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PRDX1 Protein
A cDNA sequence encoding the PRDX1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10837-100ug
Additional Details : Weight : 0.01000kg
Specifications
18477 | |
100 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is >2,500 pmol/min/ug. Enzymatic activity is defined as the amount of hydroperoxide that 1ug of enzyme can reduce at 25C for minute. | |
Mouse | |
His | |
PRDX1 protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
PRDX1 Protein | |
Research Use Only | |
Prdx1 | |
Recombinant Protein | |
Baculovirus | |
MSSGNAKIGY PAPNFKATAV MPDGQFKDIS LSEYKGKYVV FFFYPLDFTF VCPTEIIAFS DRADEFKKLN CQVIGASVDS HFCHLAWINT PKKQGGLGPM NIPLISDPKR TIAQDYGVLK ADEGISFRGL FIIDDKGILR QITINDLPVG RSVDEIIRLV QAFQFTDKHG EVCPAGWKPG SDTIKPDVNK SKEYFSKQKL EHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |