missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PDGF-B Protein
Recombinant Protein
Brand: enQuireBio™ QP5465-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_035187.2 | |
PDGF-B Protein | |
Research Use Only | |
PDGFB | |
Recombinant Protein | |
E. coli | |
MSLGSLAAAE PAVIAECKTR TEVFQISRNL IDRTNANFLV WPPCVEVQRC SGCCNNRNVQ CRASQVQMRP VQVRKIEIVR KKPIFKKATV TLEDHLACKC ETIVTPRPVT | |
Greater than 95% as determined by SDS-PAGE. |
18591 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
3T3 cell proliferation significantly induced by less than 2 ng/ml of Mouse PDGF-B. | |
Mouse | |
Untagged | |
10 mM sodium citrate, pH 3.0 |