missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse PDGF-B Protein
Recombinant Protein
£80.80 - £3965.00
Specifications
Accession Number | NP_035187.2 |
---|---|
Gene ID (Entrez) | 18591 |
Name | PDGF-B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15936189
|
enQuireBio™
QP5465-2UG |
2 μg |
£80.80
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15916189
|
enQuireBio™
QP5465-10UG |
10 μg |
£191.00
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15906189
|
enQuireBio™
QP5465-100UG |
100 μg |
£525.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946189
|
enQuireBio™
QP5465-500UG |
500 μg |
£2425.00
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926189
|
enQuireBio™
QP5465-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_035187.2 | |
PDGF-B Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
3T3 cell proliferation significantly induced by less than 2 ng/ml of Mouse PDGF-B. | |
Mouse | |
Untagged | |
10 mM sodium citrate, pH 3.0 |
18591 | |
Research Use Only | |
PDGFB | |
Recombinant Protein | |
E. coli | |
MSLGSLAAAE PAVIAECKTR TEVFQISRNL IDRTNANFLV WPPCVEVQRC SGCCNNRNVQ CRASQVQMRP VQVRKIEIVR KKPIFKKATV TLEDHLACKC ETIVTPRPVT | |
Greater than 95% as determined by SDS-PAGE. |