Learn More
enQuireBio™ Recombinant Mouse PDGF-B Protein
Recombinant Protein
Brand:  enQuireBio™ QP5465-10ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15916189
  • £191.00 / 10µg
Estimated Shipment: 13-06-2024
to see stock.
Specifications
NP_035187.2
PDGF-B Protein
Research Use Only
PDGFB
Recombinant Protein
E. coli
MSLGSLAAAE PAVIAECKTR TEVFQISRNL IDRTNANFLV WPPCVEVQRC SGCCNNRNVQ CRASQVQMRP VQVRKIEIVR KKPIFKKATV TLEDHLACKC ETIVTPRPVT
Greater than 95% as determined by SDS-PAGE.
18591
10 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
3T3 cell proliferation significantly induced by less than 2 ng/ml of Mouse PDGF-B.
Mouse
Untagged
10 mM sodium citrate, pH 3.0