missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Nidogen-1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Nidogen-1, was expressed in the hosts and tags indicated.
£272.22 - £2965.00
Specifications
Accession Number | NP_035047.2 |
---|---|
Gene ID (Entrez) | 18073 |
Name | Nidogen-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16053190
|
enQuireBio™
QP6420-YE-100UG |
100 μg |
£700.00
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16073190
|
enQuireBio™
QP6420-YE-1MG |
1 mg |
£2965.00
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063190
|
enQuireBio™
QP6420-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16083190
|
enQuireBio™
QP6420-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093190
|
enQuireBio™
QP6420-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003200
|
enQuireBio™
QP6420-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_035047.2 | |
Nidogen-1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
18073 | |
Research Use Only | |
Nid1 | |
Recombinant Protein | |
Yeast | |
SPQRVNGKVK GRIFVGSSQV PVVFENTDLH SYVVMNHGRS YTAISTIPET VGYSLLPLAP IGGIIGWMFA VEQDGFKNGF SITGGEFTRQ AEVTFLGHPG KLVLKQQFSG IDEHGHLTIS TELEGRVPQI PYGASVHIEP YTELYHYSSS VITSSSTREY TVMEPDQDGA APSHTHIYQW RQTITFQECA HDDARPALPS TQQLSVDSVF VLYNKEERIL RYALSNSIGP VRDGSPDA | |
Greater than 90% as determined by SDS-PAGE. |