missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Murinoglobulin-1 Protein
A DNA sequence encoding the Escherichia coli CFA/I fimbrial subunit E, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7135-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_032671.2 | |
Murinoglobulin-1 Protein | |
Research Use Only | |
Mug1 | |
Recombinant Protein | |
E. coli | |
TPEISWSLRT TLSKRPEEPP RKDPSSNDPL TETIRKYFPE TWVWDIVTVN STGLAEVEMT VPDTITEWKA GALCLSNDTG LGLSSVVPLQ AFKPFFVEVS LPYSVVRGEA FMLKATVMNY LPTSMQMSVQ LEASPDFTAV PVGDDQDSYC LSANGRHTSS WLVTPKSLGN VNFSVSAEAQ QSSEPCGSEV ATVPETGRKD TVVKVLIVEP E | |
Greater than 90% as determined by SDS-PAGE. |
17836 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |