missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Murinoglobulin-1 Protein
A DNA sequence encoding the Escherichia coli CFA/I fimbrial subunit E, was expressed in the hosts and tags indicated.
£265.00 - £2725.00
Specifications
Accession Number | NP_032671.2 |
---|---|
Gene ID (Entrez) | 17836 |
Name | Murinoglobulin-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16017480
|
enQuireBio™
QP7135-10UG |
10 μg |
£265.00
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057480
|
enQuireBio™
QP7135-50UG |
50 μg |
£375.00
50µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16007480
|
enQuireBio™
QP7135-100UG |
100 μg |
£625.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16037480
|
enQuireBio™
QP7135-200UG |
200 μg |
£1010.00
200µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047480
|
enQuireBio™
QP7135-500UG |
500 μg |
£1715.00
500µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027480
|
enQuireBio™
QP7135-1MG |
1 mg |
£2725.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_032671.2 | |
Murinoglobulin-1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
17836 | |
Research Use Only | |
Mug1 | |
Recombinant Protein | |
E. coli | |
TPEISWSLRT TLSKRPEEPP RKDPSSNDPL TETIRKYFPE TWVWDIVTVN STGLAEVEMT VPDTITEWKA GALCLSNDTG LGLSSVVPLQ AFKPFFVEVS LPYSVVRGEA FMLKATVMNY LPTSMQMSVQ LEASPDFTAV PVGDDQDSYC LSANGRHTSS WLVTPKSLGN VNFSVSAEAQ QSSEPCGSEV ATVPETGRKD TVVKVLIVEP E | |
Greater than 90% as determined by SDS-PAGE. |