missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Murinoglobulin-1 Protein

A DNA sequence encoding the Escherichia coli CFA/I fimbrial subunit E, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7135-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16007480

  • £625.00 / 100µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_032671.2
Murinoglobulin-1 Protein
Research Use Only
Mug1
Recombinant Protein
E. coli
TPEISWSLRT TLSKRPEEPP RKDPSSNDPL TETIRKYFPE TWVWDIVTVN STGLAEVEMT VPDTITEWKA GALCLSNDTG LGLSSVVPLQ AFKPFFVEVS LPYSVVRGEA FMLKATVMNY LPTSMQMSVQ LEASPDFTAV PVGDDQDSYC LSANGRHTSS WLVTPKSLGN VNFSVSAEAQ QSSEPCGSEV ATVPETGRKD TVVKVLIVEP E
Greater than 90% as determined by SDS-PAGE.
17836
100 μg
Not determined.
Partial (see sequence information for more details).
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers