missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse MESDC2 Protein

A cDNA sequence encoding the MESDC2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12681-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15941209

  • £99.15 / 2µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

67943
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSADTPGEA TPPPRKKKDI RDYNDADMAR LLEQWEKDDD IEEGDLPEHK RPSAPIDFSK LDPGKPESIL KMTKKGKTLM MFVTVSGNPT EKETEEITSL WQGSLFNANY DVQRFIVGSD RAIFMLRDGS YAWEIKDFLV SQDRCAEVTL EGQMYPGKGG GSKEKNKTKP EKAKKKEGDP KPRASKEDNR AGSRREDL
Greater than 90.0% as determined by SDS-PAGE.
MESDC2 Protein
Research Use Only
Mesd
Mouse
His
MESDC2 protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers