missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse MESDC2 Protein
A cDNA sequence encoding the MESDC2 was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Gene ID (Entrez) | 67943 |
---|---|
Name | MESDC2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | Mesd |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15941209
|
enQuireBio™
QP12681-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931209
|
enQuireBio™
QP12681-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15921209
|
enQuireBio™
QP12681-100UG |
100 μg |
£1320.00
100µg |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
67943 | |
Research Use Only | |
Mesd | |
Mouse | |
His | |
MESDC2 protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
MESDC2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSADTPGEA TPPPRKKKDI RDYNDADMAR LLEQWEKDDD IEEGDLPEHK RPSAPIDFSK LDPGKPESIL KMTKKGKTLM MFVTVSGNPT EKETEEITSL WQGSLFNANY DVQRFIVGSD RAIFMLRDGS YAWEIKDFLV SQDRCAEVTL EGQMYPGKGG GSKEKNKTKP EKAKKKEGDP KPRASKEDNR AGSRREDL | |
Greater than 90.0% as determined by SDS-PAGE. |