missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Mannan-binding lectin serine protease 2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Mannan-binding lectin serine protease 2, was expressed in the hosts and tags indicated.
£476.00 - £1320.00
Specifications
Accession Number | NP_001003893.1 |
---|---|
Gene ID (Entrez) | 17175 |
Name | Mannan-binding lectin serine protease 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952818
|
enQuireBio™
QP10059-BA-20UG |
20 μg |
£476.00
20µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942818
|
enQuireBio™
QP10059-BA-100UG |
100 μg |
£1320.00
100µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15962818
|
enQuireBio™
QP10059-BA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001003893.1 | |
Mannan-binding lectin serine protease 2 Protein | |
Not determined. | |
Extracellular Domain | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
17175 | |
Research Use Only | |
Masp2 | |
Recombinant Protein | |
Baculovirus | |
SKWPEPVFGR LVSPGFPEKY ADHQDRSWTL TAPPGYRLRL YFTHFDLELS YRCEYDFVKL SSGTKVLATL CGQESTDTEQ APGNDTFYSL GPSLKVTFHS DYSNEKPFTG FEAFYAAEDV DECRVSLGDS VPCDHYCHNY LGGYYCSCRA GYVLHQNKHT CSALCSGQVF TGRSGYLSSP EYPQPYPKLS SCTYSIRLED GFSVILDFVE SFDVETHPEA QCPYDSLKIQ TDKGEHGPFC GKTLPPRIET DSHKVTITFA TDESGNHTGW KIHYTSTARP CPDPTAPPNG SISPVQAIYV LKDRFSVFCK TGFELLQGSV PLKSFTAVCQ KDGSWDRPMP ECSIIDCGPP DDLPNGHVDY ITGPEVTTYK AVIQYSCEET FYTMSSNGKY VCEADGFWTS SKGEKLPPVC EPVCGLSTHT IGGRIVGGQP AKPGDFPWQV LLLGQTTAAA GALIHDNWVL TAAHAVYEKR MAASSLNIRM GILKRLSPHY TQAWPEEIFI HEGYTHGAGF DNDIALIKLK NKVTINGSIM PVCLPRKEAA SLMRTDFTGT VAGWGLTQKG LLARNLMFVD IPIADHQKCT AVYEKLYPGV RVSANMLCAG LETGGKDSCR GDSGGALVFL DNETQRWFVG GIVSWGSINC GAADQYGVYT KVINYIPWIE NIISNF | |
Greater than 90% as determined by SDS-PAGE. |