missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Kremen-1 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
£345.56
Specifications
Gene ID (Entrez) | 84035 |
---|---|
Molecular Weight (g/mol) | 42.6 kDa |
Name | Kremen-1 |
Quantity | 25μg |
Source | Mouse myeloma cell line,NS0-derived mouse Kremen-1 protein Ala20-Gly395,with a C-terminal 10-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18197180
|
R&D Systems™
1647-KR-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£345.56
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18329447
|
R&D Systems™
1647-KR-025/CF |
£345.56
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse Kremen-1 Protein is derived from NS0. The Recombinant Mouse Kremen-1 Protein has been validated for the following applications: Binding Activity.Specifications
84035 | |
Kremen-1 | |
Mouse myeloma cell line,NS0-derived mouse Kremen-1 protein Ala20-Gly395,with a C-terminal 10-His tag | |
Monomer | |
Recombinant |
42.6 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |