missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Kremen-1 Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 1647-KR-025/CF

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18329447

  • £345.56 / 25µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse Kremen-1 Protein is derived from NS0. The Recombinant Mouse Kremen-1 Protein has been validated for the following applications: Binding Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
42.6 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
84035
Kremen-1
Mouse myeloma cell line,NS0-derived mouse Kremen-1 protein Ala20-Gly395,with a C-terminal 10-His tag
Monomer
Recombinant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers