missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IGF1 Protein
A cDNA sequence encoding the IGF1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10681-50ug
This item is not returnable.
View return policy
Specifications
IGF1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGKISSLPTQ LFKICLCDFL KIKIHIMSSS HLFYLALCLL TFTSSTTAGP ETLCGAELVD ALQFVCGPRG FYFNKPTGYG SSIRRAPQTG IVDECCFRSC DLRRLEMYCA PLKPTKAARS IRAQRHTDMP KTQKEVHLKN TSRGSAGNKT YRM | |
Greater than 98.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Mouse | |
Untagged | |
The protein was lyophilized with no additives. |