missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse IGF1 Protein
A cDNA sequence encoding the IGF1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £1395.00
Specifications
Name | IGF1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Product Type | Recombinant Protein |
Cross Reactivity | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944638
|
enQuireBio™
QP10681-10UG |
10 μg |
£99.15
10µg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964638
|
enQuireBio™
QP10681-50UG |
50 μg |
£155.00
50µg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954638
|
enQuireBio™
QP10681-1MG |
1 mg |
£1395.00
1mg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
IGF1 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Mouse | |
Untagged | |
The protein was lyophilized with no additives. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGKISSLPTQ LFKICLCDFL KIKIHIMSSS HLFYLALCLL TFTSSTTAGP ETLCGAELVD ALQFVCGPRG FYFNKPTGYG SSIRRAPQTG IVDECCFRSC DLRRLEMYCA PLKPTKAARS IRAQRHTDMP KTQKEVHLKN TSRGSAGNKT YRM | |
Greater than 98.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |