missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse FKBP1A Protein
A cDNA sequence encoding the FKBP1A was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Gene ID (Entrez) | 14225 |
---|---|
Name | FKBP1A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | Fkbp1a |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15978688
|
enQuireBio™
QP11885-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968688
|
enQuireBio™
QP11885-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958688
|
enQuireBio™
QP11885-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
14225 | |
Research Use Only | |
Fkbp1a | |
Mouse | |
His | |
The FKBP1A solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT. |
FKBP1A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMGVQVE TISPGDGRTF PKRGQTCVVH YTGMLEDGKK FDSSRDRNKP FKFTLGKQEV IRGWEEGVAQ MSVGQRAKLI ISSDYAYGAT GHPGIIPPHA TLVFDVELLK LE | |
Greater than 95.0% as determined by SDS-PAGE. |