missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse FKBP1A Protein

A cDNA sequence encoding the FKBP1A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11885-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15978688

  • £99.15 / 5µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

14225
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMGVQVE TISPGDGRTF PKRGQTCVVH YTGMLEDGKK FDSSRDRNKP FKFTLGKQEV IRGWEEGVAQ MSVGQRAKLI ISSDYAYGAT GHPGIIPPHA TLVFDVELLK LE
Greater than 95.0% as determined by SDS-PAGE.
FKBP1A Protein
Research Use Only
Fkbp1a
Mouse
His
The FKBP1A solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers