missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Epididymal-specific lipocalin-5 Protein
A DNA sequence encoding the Mus musculus (Mouse) Epididymal-specific lipocalin-5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7423-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001036095.1 | |
Epididymal-specific lipocalin-5 Protein | |
Research Use Only | |
Lcn5 | |
Recombinant Protein | |
E. coli | |
TEAAVVKDFD VNKFLGFWYE IALASKMGAY GLAHKEEKMG AMVVELKENL LALTTTYYNE GHCVLEKVAA TQVDGSAKYK VTRISGEKEV VVVATDYMTY TVIDITSLVA GAVHRAMKLY SRSLDNNGEA LNNFQKIALK HGFSETDIHI LKHDLTCVNA LQSGQI | |
Greater than 90% as determined by SDS-PAGE. |
13863 | |
10 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |