missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Epididymal-specific lipocalin-5 Protein
A DNA sequence encoding the Mus musculus (Mouse) Epididymal-specific lipocalin-5, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_001036095.1 |
---|---|
Gene ID (Entrez) | 13863 |
Name | Epididymal-specific lipocalin-5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048790
|
enQuireBio™
QP7423-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16068790
|
enQuireBio™
QP7423-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058790
|
enQuireBio™
QP7423-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078790
|
enQuireBio™
QP7423-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16088790
|
enQuireBio™
QP7423-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098790
|
enQuireBio™
QP7423-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001036095.1 | |
Epididymal-specific lipocalin-5 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
13863 | |
Research Use Only | |
Lcn5 | |
Recombinant Protein | |
E. coli | |
TEAAVVKDFD VNKFLGFWYE IALASKMGAY GLAHKEEKMG AMVVELKENL LALTTTYYNE GHCVLEKVAA TQVDGSAKYK VTRISGEKEV VVVATDYMTY TVIDITSLVA GAVHRAMKLY SRSLDNNGEA LNNFQKIALK HGFSETDIHI LKHDLTCVNA LQSGQI | |
Greater than 90% as determined by SDS-PAGE. |