missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse EDA-A1/Ectodysplasin A1 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
£345.56
Specifications
Gene ID (Entrez) | 13607 |
---|---|
Molecular Weight (g/mol) | 23 kDa |
Name | EDA-A1/Ectodysplasin A1 |
Quantity | 10μg |
Source | Mouse myeloma cell line,NS0-derived mouse EDA-A1/Ectodysplasin A1 protein Ala179-Ser391,with an N-terminal 6-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18328777
|
R&D Systems™
191-ED-010/CF |
£345.56
10µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18178140
|
R&D Systems™
191-ED-010 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£345.56
10µg |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse EDA-A1/Ectodysplasin A1 Protein is derived from NS0. The Recombinant Mouse EDA-A1/Ectodysplasin A1 Protein has been validated for the following applications: Binding Activity.Specifications
13607 | |
EDA-A1/Ectodysplasin A1 | |
Mouse myeloma cell line,NS0-derived mouse EDA-A1/Ectodysplasin A1 protein Ala179-Ser391,with an N-terminal 6-His tag | |
Homotrimer | |
Recombinant |
23 kDa | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |