missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse EDA-A1/Ectodysplasin A1 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand: R&D Systems™ 191-ED-010
Additional Details : Weight : 0.00970kg
Description
The Recombinant Mouse EDA-A1/Ectodysplasin A1 Protein is derived from NS0. The Recombinant Mouse EDA-A1/Ectodysplasin A1 Protein has been validated for the following applications: Binding Activity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
23 kDa | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
13607 | |
EDA-A1/Ectodysplasin A1 | |
Mouse myeloma cell line,NS0-derived mouse EDA-A1/Ectodysplasin A1 protein Ala179-Ser391,with an N-terminal 6-His tag | |
Homotrimer | |
Recombinant |