missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse DLL4 Protein

A cDNA sequence encoding the DLL4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11670-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15937988

  • £155.00 / 10µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

DLL4 Protein
Research Use Only
Recombinant Protein
Baculovirus
GSGIFQLRLQ EFVNQRGMLA NGQSCEPGCR TFFRICLKHF QATFSEGPCT FGNVSTPVLG TNSFVVRDKN SGSGRNPLQL PFNFTWPGTF SLNIQAWHTP GDDLRPETSP GNSLISQIII QGSLAVGKIW RTDEQNDTLT RLSYSYRVIC SDNYYGESCS RLCKKRDDHF GHYECQPDGS LSCLPGWTGK YCDQPICLSG CHEQNGYCSK PDECICRPGW QGRLCNECIP HNGCRHGTCS IPWQCACDEG WGGLFCDQDL NYCTHHSPCK NGSTCSNSGP KGYTCTCLPG YTGEHCELGL SKCASNPCRN GGSCKDQENS YHCLCPPGYY GQHCEHSTLT CADSPCFNGG SCRERNQGSS YACECPPNFT GSNCEKKVDR CTSNPCANGG QCQNRGPSRT CRCRPGFTGT HCELHISDCA RSPCAHGGTC HDLENGPVCT CPAGFSGRRC EVRITHDACA SGPCFNGATC YTGLSPNNFV CNCPYGFVGS RCEFPVGLPP SFPWVAHHHH HH
Greater than 95.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Mouse
His
DLL4 protein solution (1 mg/ml) contains Phosphate buffered saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers