missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse DLL4 Protein
A cDNA sequence encoding the DLL4 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | DLL4 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Mouse |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15957988
|
enQuireBio™
QP11670-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937988
|
enQuireBio™
QP11670-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15947988
|
enQuireBio™
QP11670-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
DLL4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
DLL4 protein solution (1 mg/ml) contains Phosphate buffered saline (pH 7.4) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
GSGIFQLRLQ EFVNQRGMLA NGQSCEPGCR TFFRICLKHF QATFSEGPCT FGNVSTPVLG TNSFVVRDKN SGSGRNPLQL PFNFTWPGTF SLNIQAWHTP GDDLRPETSP GNSLISQIII QGSLAVGKIW RTDEQNDTLT RLSYSYRVIC SDNYYGESCS RLCKKRDDHF GHYECQPDGS LSCLPGWTGK YCDQPICLSG CHEQNGYCSK PDECICRPGW QGRLCNECIP HNGCRHGTCS IPWQCACDEG WGGLFCDQDL NYCTHHSPCK NGSTCSNSGP KGYTCTCLPG YTGEHCELGL SKCASNPCRN GGSCKDQENS YHCLCPPGYY GQHCEHSTLT CADSPCFNGG SCRERNQGSS YACECPPNFT GSNCEKKVDR CTSNPCANGG QCQNRGPSRT CRCRPGFTGT HCELHISDCA RSPCAHGGTC HDLENGPVCT CPAGFSGRRC EVRITHDACA SGPCFNGATC YTGLSPNNFV CNCPYGFVGS RCEFPVGLPP SFPWVAHHHH HH | |
Greater than 95.0% as determined by SDS-PAGE. |