missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRP11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24688
This item is not returnable.
View return policy
Description
NLRP11 Polyclonal specifically detects NLRP11 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| NLRP11 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CLR19.6, NACHT, leucine rich repeat and PYD containing 11, NALP11NACHT, LRR and PYD domains-containing protein 11, NLR family, pyrin domain containing 11, NOD17PYPAF7, Nucleotide-binding oligomerization domain protein 17, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 11, PAAD- and NACHT-containing protein 10, PAAD- and NACHT-containing protein 10B, PAAD-and NACHT domain-containing protein 10, PAN10FLJ26273, PYPAF6NACHT, LRR and PYD containing protein 11, PYRIN-containing APAF1-like protein 6 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 204801 | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Immunohistochemistry-Paraffin 5 ug/ml | |
| P59045 | |
| NLRP11 | |
| A portion of amino acids 850-900 of human NALP11 was used as the immunogen. | |
| 0.1 mL | |
| Primary | |
| This antibody will detect the approximately 300 kD full-length Notch1form and the approximately 100 kD cleaved Notch1 form of the protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido