missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRP11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £301.00
Specifications
| Antigen | NLRP11 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18140512
|
Novus Biologicals
NBP2-24688SS |
0.025 mL |
£156.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18197481
|
Novus Biologicals
NBP2-24688 |
0.1 mL |
£301.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NLRP11 Polyclonal specifically detects NLRP11 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NLRP11 | |
| Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| CLR19.6, NACHT, leucine rich repeat and PYD containing 11, NALP11NACHT, LRR and PYD domains-containing protein 11, NLR family, pyrin domain containing 11, NOD17PYPAF7, Nucleotide-binding oligomerization domain protein 17, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 11, PAAD- and NACHT-containing protein 10, PAAD- and NACHT-containing protein 10B, PAAD-and NACHT domain-containing protein 10, PAN10FLJ26273, PYPAF6NACHT, LRR and PYD containing protein 11, PYRIN-containing APAF1-like protein 6 | |
| NLRP11 | |
| IgG | |
| Protein A purified | |
| This antibody will detect the approximately 300 kD full-length Notch1form and the approximately 100 kD cleaved Notch1 form of the protein. |
| 0.5 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P59045 | |
| 204801 | |
| A portion of amino acids 850-900 of human NALP11 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title