missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Cerebellin-3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Cerebellin-3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10025-iv-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_062794.1 | |
Cerebellin-3 Protein | |
Research Use Only | |
Cbln3 | |
Recombinant Protein | |
E. coli | |
QEGSEPVLLE GECLVVCEPG RPTAGGPGGA ALGEAPPGRV AFAAVRSHHH EPAGETGNGT SGAIYFDQVL VNEGEGFDRT SGCFVAPVRG VYSFRFHVVK VYNRQTVQVS LMLNTWPVIS AFANDPDVTR EAATSSVLLP LDPGDRVSLR LRRGNLLGGW KYSSFSGFLI FPL | |
Greater than 80% as determined by SDS-PAGE. |
56410 | |
50 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |