missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Cerebellin-3 Protein
A DNA sequence encoding the Mus musculus (Mouse) Cerebellin-3, was expressed in the hosts and tags indicated.
£428.07 - £1240.00
Specifications
Accession Number | NP_062794.1 |
---|---|
Gene ID (Entrez) | 56410 |
Name | Cerebellin-3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15962768
|
enQuireBio™
QP10025-IV-100UG |
100 μg |
£1240.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15982768
|
enQuireBio™
QP10025-IV-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15972768
|
enQuireBio™
QP10025-IV-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_062794.1 | |
Cerebellin-3 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
56410 | |
Research Use Only | |
Cbln3 | |
Recombinant Protein | |
E. coli | |
QEGSEPVLLE GECLVVCEPG RPTAGGPGGA ALGEAPPGRV AFAAVRSHHH EPAGETGNGT SGAIYFDQVL VNEGEGFDRT SGCFVAPVRG VYSFRFHVVK VYNRQTVQVS LMLNTWPVIS AFANDPDVTR EAATSSVLLP LDPGDRVSLR LRRGNLLGGW KYSSFSGFLI FPL | |
Greater than 80% as determined by SDS-PAGE. |