missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD80 Protein
A cDNA sequence encoding the CD80 was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Gene ID (Entrez) | 12519 |
---|---|
Name | CD80 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | Cd80 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976968
|
enQuireBio™
QP11349-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956968
|
enQuireBio™
QP11349-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966968
|
enQuireBio™
QP11349-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
12519 | |
Research Use Only | |
Cd80 | |
Mouse | |
His | |
CD80 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
CD80 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
DVDEQLSKSV KDKVLLPCRY NSPHEDESED RIYWQKHDKV VLSVIAGKLK VWPEYKNRTL YDNTTYSLII LGLVLSDRGT YSCVVQKKER GTYEVKHLAL VKLSIKADFS TPNITESGNP SADTKRITCF ASGGFPKPRF SWLENGRELP GINTTISQDP ESELYTISSQ LDFNTTRNHT IKCLIKYGDA HVSEDFTWEK PPEDPPDSKN HHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |