missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD80 Protein
A cDNA sequence encoding the CD80 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11349-2ug
Additional Details : Weight : 0.01000kg
Specifications
12519 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
DVDEQLSKSV KDKVLLPCRY NSPHEDESED RIYWQKHDKV VLSVIAGKLK VWPEYKNRTL YDNTTYSLII LGLVLSDRGT YSCVVQKKER GTYEVKHLAL VKLSIKADFS TPNITESGNP SADTKRITCF ASGGFPKPRF SWLENGRELP GINTTISQDP ESELYTISSQ LDFNTTRNHT IKCLIKYGDA HVSEDFTWEK PPEDPPDSKN HHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
CD80 Protein | |
Research Use Only | |
Cd80 | |
Mouse | |
His | |
CD80 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |