missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse CD5L Protein

A cDNA sequence encoding the CD5L was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11342-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15966938

  • £99.15 / 2µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

11801
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Baculovirus
ESPTKVQLVG GAHRCEGRVE VEHNGQWGTV CDDGWDRRDV AVVCRELNCG AVIQTPRGAS YQPPASEQRV LIQGVDCNGT EDTLAQCELN YDVFDCSHEE DAGAQCENPD SDLLFIPEDV RLVDGPGHCQ GRVEVLHQSQ WSTVCKAGWN LQVSKVVCRQ LGCGRALLTY GSCNKSTQGK GPIWMGKMSC SGQEANLRSC LLSRLENNCT HGEDTWMECE DPFELKLVGG DTPCSGRLEV LHKGSWGSVC DDNWGEKEDQ VVCKQLGCGK SLHPSPKTRK IYGPGAGRIW LDDVNCSGKE QSLEFCRHRL WGYHDCTHKE DVEVICTDFD VLEHHHHHH
Greater than 95.0% as determined by SDS-PAGE.
CD5L Protein
Research Use Only
Cd5l
Mouse
His
CD5L protein solution (0.25 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers