missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD5L Protein
A cDNA sequence encoding the CD5L was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Gene ID (Entrez) | 11801 |
---|---|
Name | CD5L Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | Cd5l |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15966938
|
enQuireBio™
QP11342-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946938
|
enQuireBio™
QP11342-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956938
|
enQuireBio™
QP11342-1MG |
1 mg |
£5730.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
11801 | |
Research Use Only | |
Cd5l | |
Mouse | |
His | |
CD5L protein solution (0.25 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |
CD5L Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ESPTKVQLVG GAHRCEGRVE VEHNGQWGTV CDDGWDRRDV AVVCRELNCG AVIQTPRGAS YQPPASEQRV LIQGVDCNGT EDTLAQCELN YDVFDCSHEE DAGAQCENPD SDLLFIPEDV RLVDGPGHCQ GRVEVLHQSQ WSTVCKAGWN LQVSKVVCRQ LGCGRALLTY GSCNKSTQGK GPIWMGKMSC SGQEANLRSC LLSRLENNCT HGEDTWMECE DPFELKLVGG DTPCSGRLEV LHKGSWGSVC DDNWGEKEDQ VVCKQLGCGK SLHPSPKTRK IYGPGAGRIW LDDVNCSGKE QSLEFCRHRL WGYHDCTHKE DVEVICTDFD VLEHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |