missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CD5L Protein
A cDNA sequence encoding the CD5L was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11342-10ug
Additional Details : Weight : 0.01000kg
Specifications
11801 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ESPTKVQLVG GAHRCEGRVE VEHNGQWGTV CDDGWDRRDV AVVCRELNCG AVIQTPRGAS YQPPASEQRV LIQGVDCNGT EDTLAQCELN YDVFDCSHEE DAGAQCENPD SDLLFIPEDV RLVDGPGHCQ GRVEVLHQSQ WSTVCKAGWN LQVSKVVCRQ LGCGRALLTY GSCNKSTQGK GPIWMGKMSC SGQEANLRSC LLSRLENNCT HGEDTWMECE DPFELKLVGG DTPCSGRLEV LHKGSWGSVC DDNWGEKEDQ VVCKQLGCGK SLHPSPKTRK IYGPGAGRIW LDDVNCSGKE QSLEFCRHRL WGYHDCTHKE DVEVICTDFD VLEHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
CD5L Protein | |
Research Use Only | |
Cd5l | |
Mouse | |
His | |
CD5L protein solution (0.25 mg/ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |