missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Arginase 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Arginase 1, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_031508.1 |
---|---|
Gene ID (Entrez) | 11846 |
Name | Arginase 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16050431
|
enQuireBio™
QP7724-YE-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16070431
|
enQuireBio™
QP7724-YE-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060431
|
enQuireBio™
QP7724-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080431
|
enQuireBio™
QP7724-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16090431
|
enQuireBio™
QP7724-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16000441
|
enQuireBio™
QP7724-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_031508.1 | |
Arginase 1 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
11846 | |
Research Use Only | |
ARG1 | |
Recombinant Protein | |
Yeast | |
MSSKPKSLEI IGAPFSKGQP RGGVEKGPAA LRKAGLLEKL KETEYDVRDH GDLAFVDVPN DSSFQIVKNP RSVGKANEEL AGVVAEVQKN GRVSVVLGGD HSLAVGSISG HARVHPDLCV IWVDAHTDIN TPLTTSSGNL HGQPVSFLLK ELKGKFPDVP GFSWVTPCIS AKDIVYIGLR DVDPGEHYII KTLGIKYFSM TEVDKLGIGK VMEETFSYLL GRKKRPIHLS FDVDGLDPAF TPATGTPVLG GLSYREGLYI TEEIYKTGLL SGLDIMEVNP TLGKTAEEVK STVNTAVALT LACFGTQREG NHKPGTDYLK PPK | |
Greater than 90% as determined by SDS-PAGE. |