missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Adamts5 Protein
A DNA sequence encoding the Mus musculus (Mouse) Adamts5, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | NP_035912.2 |
---|---|
Gene ID (Entrez) | 23794 |
Name | Adamts5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003411
|
enQuireBio™
QP8228-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16023411
|
enQuireBio™
QP8228-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16013411
|
enQuireBio™
QP8228-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033411
|
enQuireBio™
QP8228-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043411
|
enQuireBio™
QP8228-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053411
|
enQuireBio™
QP8228-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_035912.2 | |
Adamts5 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
23794 | |
Research Use Only | |
Adamts5 | |
Recombinant Protein | |
E. coli | |
SISRARQVEL LLVADSSMAR MYGRGLQHYL LTLASIANRL YSHASIENHI RLAVVKVVVL TDKDTSLEVS KNAATTLKNF CKWQHQHNQL GDDHEEHYDA AILFTREDLC GHHSCDTLGM ADVGTICSPE RSCAVIEDDG LHAAFTVAHE IGHLLGLSHD DSKFCEENFG TTEDKRLMSS ILTSIDASKP WSKCTSATIT EFLDDGHGNC LLDLPRKQIL GPEELPGQTY DATQQCNLTF GPEYSVCPGM DVCARLWCAV VRQGQMVCLT KKLPAVEGTP CGKGRVCLQG KCVDKTKKKY YSTSSHGNWG SWGPWGQCSR SCGGGVQFAY RHCNNPAPRN SGRYCTGKRA IYRSCSVTPC PPNGKSFRHE QCEAKNGYQS DAKGVKTFVE WVPKYAGVLP ADVCKLTCRA KGTGYYVVFS PKVTDGTECR PYSNSVCVRG RCVRTGCDGI IGSKLQYDKC GVCGGDNSSC TKIIGTFNKK SKGYTDVVRI PEGATHIKVR QFKAKDQTRF TAYLALKKKT GEYLINGKYM ISTSETIIDI NGTVMNYSGW SHRDDFLHGM GYSATKEILI VQILATDPTK ALDVRYSFFV PKKTTQKVNS VISHGSNKVG PHSTQLQWVT GPWLACSRTC DTGWHTRTVQ CQDGNRKLAK GCLLSQRPSA FKQCLLKKC | |
Greater than 90% as determined by SDS-PAGE. |